Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromo-2,5-dimethoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D396625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2674-34-2
Molecular Formula C8H8Br2O2
Molecular Weight (g/mol) 295.958
MDL Number MFCD00025694
InChI Key CHCLRVOURKGRSW-UHFFFAOYSA-N
Synonym 2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene
PubChem CID 231240
IUPAC Name 1,4-dibromo-2,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1Br)OC)Br

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 25 g
Formula Weight 295.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-dimethoxybenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.