Learn More
Cole-Parmer Partlow 51100011 2 Chnl Rec Nema 3 Case, 120 volt
Supplier: Cole-Parmer 4900187
These recorders are designed with the latest innovation in recording technology, enclosures, and functionality. Finding a place to install this recorder is easy with the compact 2.5" panel depth and short 1.3" protrusion — cutout size is the commonly-utilized 12.7" square, too! It can be panel or surface mounted.