Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ GST Activity Assay Kit (Fluorometric)
To detect the GST activity, functional study
$981.00 - $1073.00
Specifications
Content And Storage | Store the kit at -20°C. |
---|---|
Detection Method | Fluorometric |
For Use With (Application) | Functional Study |
Quantity | 1 Kit |
Type | Activity Assay Kit |
Catalog Number | Mfr. No. | Sample Type | Calibration Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sample Type | Calibration Range | Price | Quantity & Availability | ||||
89-028-099
|
Abnova™
KA0796 |
Cell Sample, Erythrocute Sample, Plasma Sample, Tissue Sample | 0.5 mU |
Each for $981.00
|
|
||||
89-101-266
|
Abnova™
KA3747 |
Cell Lysate, Serum, Tissue Lysate | Less than 0.02 mIU |
Each for $1,073.00
|
|
||||
Specifications
Store the kit at -20°C. | |
Functional Study | |
Activity Assay Kit |
Fluorometric | |
1 Kit |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title