Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Immune Complex (24C5-BNP/PROBNP), In Vitro, 0.2mg
SDP
Supplier:  Advanced ImmunoChemical, Inc. 2ICIV
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal mouse anti-immune complex (24C5-BNP/proBNP), in vitro. MAbs:Ab-BNP2ccHybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with immune complex formed by Fab-fragment of BNP-specific MAb 24C5 (Cat.# 2-BNP) and synthetic human BNP.Specificity:Human BNP and proBNP in immune complex with MAb 24C5MAb Isotypes:IgG2a for MAb Ab-BNP2ccApplications:BNP and proBNP sandwich immunoassays (detection), only as a pair to MAb 24C5.Purification:Protein A affinity chromatographyPresentation:PBS, pH 7.4, 0.09 % sodium azide (NaN3)Storage Conditions:+ 4 °COther information:More antibodies against immune complex (24C5-BNP/proBNP) under Cat.# 2-ICBNP.Material safety note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. 50-153-9867
Explore available promotions