Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Kidney Injury Molecule-1 (KIM 1), 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 2KIM10.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal Antibodies / Tumor Markers / Monoclonal mouse anti-human kidney injury molecule (KIM-1). MAbs:KIM70, KIM75Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant KIM-1 ectodomain expressed in baculovirus system.Specificity:Human KIM-1MAb Isotypes:IgG1 for MAbs KIM70, KIM75Applications:MAbs KIM70, KIM75 react with human recombinant KIM-1 in sandwich ELISA and Western blotting.Recommended pairs KIM-1 immunodetection in sandwich immunoassay (capture-detection):KIM70 – KIM75Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °CMaterial Safety Note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.