Learn More
United States Biological Corporation BACTO YEAST EXTRACT 500G
SDP
Supplier:  United States Biological Corporation Y2010500G
Encompass_Preferred
Aqueous extract of brewers yeast with an exceptionally high vitamin content. Selected for greater solubility and clarity. Free amino acid content of approximately 25%. Yeast Extract is produced from autolyzed yeast cells and is very soluble in water. It is used as an enrichment in a large number of culture media for general bacteriology and in media for sterility according to the USP. Because of its high content of carbohydrates, Yeast Extract should not be used in fermentation studies. Typical Amino Acid Profile: Ala 42%, Arg 27%, Asp 59%, Cys 5%, Glu 105%, Gly 27%, His 12%, Ile 28%, Leu 42%, Lys 45%, Met 9%, Phe 24%, Pro 21%, Ser 27%, Thr 28%, Trp 6%, Tyr 21%, Val 36% Appearance: Light yellow to medium tan, homogeneous, free flowing powder Solubility: Light yellow to amber, clear, complete pH: 6.5 � 0.5 Ash: 17% Salt: 0.5% Nitrogen (Amino): 5% Nitrogen (AN/TN): 36% Microbiological Testing: Salmonella, Standard Plate Count, Coliform (MPN/g), Viable Yeast & Mold.
Catalog No. NC9046283
Explore available promotions