Learn More
Research Products International Corp BoxTop Parafilm DIS, BL
Supplier: Research Products International Corp 249412B
BoxTop Parafilm Disp conveniently fits on box of Parafilm M to cut film quickly while protecting finger TPS from injury and sharp blades. Easy to use - place BoxTop Disp on top of a parafim box (rolls do not need to be removed) and pull sealing film through. Built-in safety blade slides and cuts film easily without using scissors. Made of durable ABS PLAS and supplied with two safety blades. Each Disp holds 2 x 2 inch roll or 1 x 4 inch roll.Conveniently fits on Parafilm box Cuts film quicklyEconomicalDimensions (inches): 4.9 x 6.8 x 3.9 HShipping Weight: 1 lbsShipping Dimensions: 5.00 x 7.00 x 4.00Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.