Learn More
Advanced ImmunoChemical, Inc. CA72-4, Source - Huma Metastatic Liver Carcinoma, 250 kIU
SDP
Supplier:  Advanced ImmunoChemical, Inc. 8CA724
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Tumor Markers / CA72-4. Source: Human metastatic liver carcinoma.Blood sample from the tissue donor was tested and found negative for HBsAg, HIV-1 and HIV-2 antibodies and HCV.Storage: -20 °C (-15 … -30 °C allowed). Applications: Immunogen, calibrator in immunoassays, in vitro studies.Presentation: PBS, pH 7.4, 0.05 % of sodium azide (NaN₃).Material safety note: This product is sold for research use only. Standard Laboratory Practices should be followed when handling this material. Product contains sodium azide as a preservative. Although the amount of sodium azide is very small appropriate care must be taken when handling this product.
Catalog No. 50-153-9854
Explore available promotions