Learn More
Chemglass Life Sciences Condenser, Graham, 530mm, 24/40 Joint, 400mm Jacket Length
Supplier: Chemglass Life Sciences CG121407
Condenser, Graham, 530mm, 24/40 Joint, 400mm Jacket Length. Condenser has a top standard taper outer and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.