Learn More
Chemglass Life Sciences Graham Condenser
SDP
Supplier:  Chemglass Life Sciences CG183030
Encompass_Preferred
Condenser for use in cooling of the scrubbing fluid. Graham coil type condenser having 1/2 Swagelok tubing for use with CG-1975-20 compression fitting. Overall length is ~350mm with effective length of 250mm. Hose connections have 9mm OD small serration and a 10mm OD large serration.
Catalog No. 50-873-387
Explore available promotions