Learn More
Hach Company Cylinder, Graduated, Mixing, Glass, 25 mL +-0.3 mL, 0.5mL divisions, glass stopper #13
Supplier:  Hach Company 189640
Encompass_Preferred
Scale is durable white ceramic enamel. Designed to meet ASTM Specification E1272, Style 2 (Class B). Includes glass stopper.
Catalog No. 50-199-4632
Explore available promotions