Learn More
Kamiya Biomedical D-Dimer Reagent, 16mL+8.5mL/PK RX
SDP
Supplier:  Kamiya Biomedical KAI090
Encompass_Preferred
K-ASSAY D-Dimer Assay Reagent. R1: 1 x 16mL (liquid), R2: 1 x 8.5mL (liquid). Assay Range: 0.5 - 30ug/mL DDU (or value of highest calibration point). The K-ASSAY D-Dimer Assay is an in vitro diagnostic reagent for the quantitative determination of cross-linked fibrin degradation products containing D-Dimer in human plasma or serum by immunoturbidimetric assay. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY D-Dimer Calibrator (Cat. No. KAI-091C). Recommended Controls: K-ASSAY Plasma Control (Cat. No. K81C-6M). This assay can be used on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
Catalog No. 50-465-387
Explore available promotions