Learn More
Perkin Elmer US LLC Demountable Quartz Injector - 1.5 mm I.D., Black/Orange
Supplier: Perkin Elmer US LLC N8152445
Quartz injectors are used for volatile organic solvent introduction and are recommended for a wide range of non-HF applications. The Injector Light Shield Disk (N8152682) is included.