Learn More
StatLab Eosin Y Stain, Phloxine B, Mastertech, 1 L
SDP
Supplier:  StatLab STE0250
Encompass_Preferred
Another flagship stain from American MasterTech, this superior Eosin Y formulation has remained popular for more than 30 years! It stains cytoplasmic structures, collagen, and basement membranes various shades of pink and is made from BSC certified dyes. Phloxine B formula stains structures with a more reddish tint for pathologists that prefer it.
Catalog No. NC1125836
Explore available promotions