Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin A Precursor Antibody, R&D Systems™
Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | Activin A |
---|---|
Clone | 132815 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Activin A Monoclonal specifically detects Activin A in Human samples. It is validated for Western Blot.Specifications
Activin A | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
INHBA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
132815 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DNA construct encoding human Activin A pro region fused to mature human Activin beta B chain | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Activin A Precursor in direct ELISAs and Western blots. Recognizes an epitope found in the human Activin A pro region that is removed when the protein is processed to its mature form. |
For Research Use Only
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title