Learn More
Caron Products And Services Humidified Incubator, 25 cu. ft., Temp & RH
Supplier: Caron Products And Services 7003252
25 ft3 (708 L) Humidified Incubator offers temperature (10C above ambient-70C) and humidity control (ambient to 98% RH). Ideal for a wide range of applications. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.