Learn More
TARGETMOL CHEMICALS INC IMIDACLOPRID-UREA 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T72077
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Imidacloprid-urea is a secondary metabolite of imidacloprid. Imidacloprid is a potent and widely used neonicotinoid with inhibitory effects on pests of cereals vegetables tea and cotton. imidacloprid-urea can occupy or block the adsorption sites of imidacloprid on the soil which may affect the fate transport and bioavailability of imidacloprid in the environment. purity: 99%
Catalog No. 50-257-9244
Explore available promotions