Learn More
BioMed Diagnostics A self-contained system for the detection of Tritrichomonas foetus in feline samples. | Detects: Tritrichomonas foetus. | Available in: 100 pack
Supplier:  BioMed Diagnostics 12071004
Encompass_Preferred
A self-contained system for the detection of Tritrichomonas foetus in feline samples. The proprietary medium is selective for the transport and growth of the trichomonas while inhibiting the growth of mold, bacteria, and yeast which could interfere with diagnosis. | Product Principle: 1.) Combine culture, result and microscopic observation in one IVD device. 2.) Highly selective prepared media in a device designed for in-vitro specimen transport and incubation. 3.) Rapid presumptive diagnosis of T. foetus in cats. 4.) Recommended as a parallel test with other POC rapid tests. 5.) Higher recovery rate of T. foetus from cats compared to Diamond media. 6.) InPouch can be placed directly on microscope stage. | Detects: T. foetus (Tritrichomonas foetus) | Time to Result: 15-30 minutes. | Available in: 100 pack
Catalog No. NC2582831
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions