Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Full Length Stoppers
Stoppers are used with laboratory glassware such as flasks and separatory funnels.
$89.40 - $136.00
Specifications
Material | Glass |
---|---|
Product Type | Solid Pennyhead Stopper |
For Use With (Equipment) | Flask and separatory funnel |
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity & Availability | |||||
K851000-3445
|
DWK Life Sciences
8510003445 |
34/45 |
Case for $96.00
|
|
|||||
K851000-2942
|
DWK Life Sciences
8510002942 |
29/42 |
Case for $89.40
|
|
|||||
K851000-4550
|
DWK Life Sciences
8510004550 |
45/50 |
Case for $136.00
|
|
|||||
Description
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Specifications
Glass | |
Solid Pennyhead Stopper | |
Flask and separatory funnel |