Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Lab Bench Vert Organizer, 4 Pocket, Double-wide
SDP

Benchtop organizer for tip boxes, bottles, etc. Made of durable PETG construction.

$167.00 - $243.00

Specifications

Autoclavable Not Autoclavable
For Use With (Application) Pipette Tip Boxes
Width (Metric) 242 mm
Product Type Organizer
Color Clear
View More Specs

Products 4
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
22-000-788
SDP
View Documents
Poltex™
4VTORGDBL
Organizer
Each for $167.00
Add to Cart
 
22-000-789
SDP
View Documents
Poltex™
4VTORGDBLPIPTRBRK
Organizer, Pipettor Bracket
Each for $216.00
Add to Cart
 
22-000-772
SDP
View Documents
Poltex™
4VTORGDBLBRK
Organizer, Pipette Aid Bracket
Each for $195.00
Add to Cart
 
22-000-764
SDP
View Documents
Poltex™
4VTORGDBLPIPTRBRKBRK
Organizer, Pipetter, Pipette Aid Brackets
Each for $243.00
Add to Cart
 
Description

Description

  • Free up valuable lab bench space by going up!
  • Highly durable, 1/4 in. thick PETG construction
  • Holds pipette tips, bottles, etc.
  • Comes with optional accessories: 4 pipettor bracket and/or pipette controller bracket
  • Rubber bumpons on base for no slip placement
Specifications

Specifications

Not Autoclavable
242 mm
Clear
Pipette Tip Boxes
Organizer
PETG
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.