Learn More
Cole-Parmer Cole-Parmer LED Fiber Optic Illuminator, Dual-Gooseneck Light Pipe; 115 VAC
Supplier:  Cole-Parmer 4172302
Encompass_Preferred
High-power white LEDs provide uniform color temperature, high light output, low heat, low power consumption, and extremely long life. Ideal for use in microscopy applications, inspection, forensics, machine vision, and assembly stations. Easily adjust the light output using the variable 10 to 100% intensity control. Built-in handle stays out of the way and allows for stacking of multiple units. Illuminator is available with a gooseneck light pipe or ring lights.
Catalog No. NC1782887
Please call customer service for current pricing. \n1-800-766-7000
Explore available promotions