Learn More
Enzo Life Sciences Matriptase-2 (human), (recombinant) (500U)
SDP
Supplier:  Enzo Life Sciences ALX2017520500
Encompass_Preferred
Alternative name: Transmembrane protease serine 6, TMPRSS6. MW: ~84kDa; a-subunit: ~56kDa; b-subunit: ~28kDa. Formulation: Liquid. In 100mM TRIS-HCl, pH 9.0, containing 150 mM sodium chloride and 20% glycerol. Source: Produced in Drosophila Schneider 2 (S2) cells. Recombinant human matriptase-2 (aa 78-811) is fused at the C-terminus to a V5-His-tag. Produced by plasmid transfection expression. Long Term Storage: -80°C. Use/Stability: Stable for at least 6 months when stored at -20°C. Recommended Assay Buffer: 100mM TRIS, pH 9.0, containing 0.5mg/ml BSA. Do not store diluted solutions. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7981
Explore available promotions