Learn More
Biotang Inc PCR Tube & Capstrip - 8strip with flat caps, natural, 250 strips/2000 tubes/case. Fit most thermal cyclers, PCR instruments, and DNA sequencers. Real-Time PCR/QPCR compatible. Dnase, Rnase, Human DNA, Pyrogen, PCR Inhibitors Free
Supplier: Biotang Inc BT37350
PCR Tube & Capstrip - 8strip with flat caps, natural, 250 strips/2000 tubes/case. Fit most thermal cyclers, PCR instruments, and DNA sequencers. Real-Time PCR/QPCR compatible. Dnase, Rnase, Human DNA, Pyrogen, PCR Inhibitors Free