Learn More
Poltex 6 Pocket PETG Serological Pipette Holder
The PETG Serological Pipette Holder is available in 3 pocket capacities, with 3 mounting options, and the option to add a pipette filler bracket to any version.
Supplier: Poltex PGSERPIP6
Description
- Serological Pipette (pipet) Holder.
- Choose from 4, 5, or 6 pocket capacity.
- Choose from powerful, interchangeable magnet mount, 3M VHB (very high bond) tape mount, or countertop placement.
- Add a pipette filler bracket to any version (bracket mounts to either side).
- Magnets and tape mount to either side.
- Break-resistant PETG construction.
- PETG has good alcohol/chemical resistance.
- Shelves are removable for easy cleaning.
Specifications
Serological Pipet Holder | |
PETG | |
Countertop |
Holder Only | |
6 Pockets |