Learn More
Ace Glass, Inc. Photochem reactor body, 1000mL, complete with spinbar, sparger, bushing and PTFE tubing
Supplier: Ace Glass, Inc. 784119
Complete photochemical reactor featuring spinbar, sparger, bushing and PTFE tubing. 1000mL reaction vessel is made of borosilicate glass to accomm ODate ACE 7854, 7856, or 7857 immersion wells. Vessel has one 14/20 angled joint for sparger tube, one 24/40 vertical joint for condenser, and one #7 Ace-Thred side arm for a thermometer. Center joint is 60/40, and the bottom of the reactor is flat to allow use of a stir magnet.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.