Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pro-Lab Diagnostics™ Prolex™ Staph Xtra Latex Kits
A rapid, sensitive method to distinguish Staphylococcus aureus and methicillin resistant Staphylococcus aureus (MRSA) from other species of Staphylococci
$319.00
Specifications
Detectable Analytes | S. aureus |
---|---|
Type | Staph Xtra Latex Kit |
Includes:
- Staph Xtra Latex Reagent: 2 x 2.5mL (PL.1080), 2 x 7.5mL (PL.1081)
- Negative Control Latex Reagent: 5.0mL (PL.1080), 2 x 7.5mL (PL.1081)
- Agglutination Cards: 20 (PL.1080), 48 (PL.1081)
- Mixing Sticks: 200 (PL.1080), 300 (PL.1081)
Catalog Number | Mfr. No. | Quantity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Price | Quantity & Availability | ||||
22-286-204
|
Pro-Lab Diagnostics™
PL.1080 |
100 Tests | 2 x 2.5mL Staph Xtra Latex reagent, 2.5mL negative control Latex reagent, 20 Agglutination cards, 200 mixing sticks |
Each for $319.00
|
|
||||
22-286-202
|
Pro-Lab Diagnostics™
PL.1081 |
300 Tests | 2 x 2.7mL Staph Xtra Latex reagent, 7.5mL negative control Latex, 48 Agglutination cards | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Rapid clear results within 20 seconds
- Uses blue latex particles (CMMP) against a white background
Specifications
S. aureus | |
Staph Xtra Latex Kit |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title