Learn More
Enzo Life Sciences Prolactin (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1940050
Encompass_Preferred
Alternative name: Luteotropic hormone , Luteotropin. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized from 1X PBS, pH 7.5. Source: Produced in HEK 293 cells. Prolactin (aa 29-227) is fused at the N-terminus to a FLAG-tag, 13 amino acid spacer, and a His-tag. Long Term Storage: -20°C.
Catalog No. 50-201-3982
Explore available promotions