Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Sample Processing Kit, blood samples

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

$318.00 - $4765.00

Specifications

Format Kit
Sample Type Blood
Product Line QuantiGene™
Product Type Sample Processing Kit
Shipping Condition Blue Ice
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
QS0110
Encompass_Preferred
View Documents
Invitrogen™
QS0110
2 Plates
Each for $318.00
Add to Cart
 
QS0111
Encompass_Preferred
View Documents
Invitrogen™
QS0111
10 Plates
Each for $1,264.00
Add to Cart
 
QS0112
Encompass_Preferred
View Documents
Invitrogen™
QS0112
5 x 10 Plates
Each for $4,765.00
Add to Cart
 
Description

Description

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your whole blood or dried blood spot sample

What you get

The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.

Specifications

Specifications

Kit
QuantiGene™
Blue Ice
Blood
Sample Processing Kit
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.