Learn More
StatLab R.B.D DECAL SOL 1GAL
SDP
Supplier:  StatLab DCR0257
Encompass_Preferred
Rapid Bone Decalcifier Decal Solution; 1 gal.; Very fast and aggressive decal solution just like R.D.O. decalcifier; Most bone marrow biopsies will decalcify in 30 min.; Biodegradable and non-corrosive
Catalog No. NC9327337
Explore available promotions