Learn More
Ace Glass, Inc. 500mL, one-piece pressure reactor with #25 center, (3)#15, (1)#7 side necks, complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies
SDP
Supplier:  Ace Glass, Inc. 6436233
Encompass_Preferred
Request Bulk Order
500mL one-piece pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Flask features #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. System is complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies. Rated 45 PSI at 100C.
Catalog No. 50-978-716
Explore available promotions