Learn More
Ace Glass, Inc. Reactor, Kriel, 5000mL complete
Supplier: Ace Glass, Inc. 696265
5000mL complete Kriel Photochemical reactor includes a five-neck tapered flask with a #50 center neck, (2) 29/42 side necks, and (2) #7 Ace-Thred side necks, quartz immersion well, glass stopper, stirring shaft, stir bearing, and stir magnet. The reactor is most useful in prep-scale photolysis, and the tapered sides of the flask assure full submersion using either 509558006 or 50975483 immersion lamps within the supplied quartz immersion well, which allows for a 20% savings in reaction time over round bottom flasks. Additionally, the flat bottom design allows for immediate stirring with a magnetic stirrer, and mechanical stirring after only being 1/3 full.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.