Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Safety Sign: AUTHORIZED PERSONNEL ONLY
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements.
Supplier: Accuform™ MADM499VS
Description
- Ideal for indoor and sheltered outdoor marking needs
- Adhere to many flat or irregular surfaces
- Flexible opaque poly with a printed matte surface
- Permanent adhesive back with peel-off liner
Specifications
OSHA 29 CFR 1910.145, ANSI Z535 | |
Polyester | |
10 x 14 in. | |
Self-adhesive | |
English | |
Safety Sign |
Non-glare | |
1 Sign | |
Pressure-sensitive | |
Rectangular | |
AUTHORIZED PERSONNEL ONLY | |
Red, White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction