Learn More
ICU Medical Anaerobic Pulsator™ Plus Arterial Blood Sampling Kit
Pulsator™ Plus syringes, with a liquid sodium heparin concentration of 23.5 I.U. per mL, allowing blood sampling with minimal risk of sample dilution. The syringes use a specially formulated electrolyte-balanced liquid sodium heparin that offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels. The syringes are available with a broad choice of product features.
Supplier: ICU Medical 4058NP2
Description
Anaerobic Pulsator Plus syringes, with a liquid sodium heparin concentration of 23.5 IU/mL, allow for clot-free blood sampling without risking sample dilution. Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels, providing consistent and accurate analysis for all parameters tested.
- Consistent and accurate readings of pH, pOC2, pO2, Ca2+, Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactateFill rapidly, to reduce fill time and minimize patient discomfort
- Pulsating effect allows for precise positioning in arterial lumen
- Syringe can be easily aspirated during a low blood pressure or code situation
- Patented mini-volume insert needle and hub has a residual liquid heparin dead space of 0.03 mL; allows liquid heparin to be thoroughly expelled from syringe and needle, leaves behind an accurate trace coating of heparin
- Needles available with different gauges, lengths and safety devices (Needle-Pro™, Point-Lok™ or Filter-Pro™)
- With or without support kit (alcohol prep pad, iodophor prep pad, two gauze pads, patient label, biohazard labeled ice bag, adhesive bandage)
Order Info
- Needles available with different gauges, lengths and safety devices (Needle-Pro™, Point-Lok™ or Filter-Pro™)
- With or without support kit (alcohol prep pad, iodophor prep pad, two gauze pads, patient label, biohazard labeled ice bag, adhesive bandage)
Specifications
22 G | |
3mL luer-lock syringe with Filter-Pro™ device, 22G x 1.5 in. needle with separate Needle-Pro™ needle safety device a mini-volume insert needle, 23.5 IU/mL concentration of liquid sodium heparin | |
200/Cs. | |
3 mL |
Allow for clot-free blood sampling without risking sample dilution, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels, providing consistent and accurate analysis for all parameters tested | |
Arterial Blood Sampling Kit | |
Polybag | |
Sterile |
Safety and Handling
ShelfLife : 3 Years
Your input is important to us. Please complete this form to provide feedback related to the content on this product.