Learn More
Bellco Glass, Inc. Stainless steel port plug fits onto stainless steel head plate for 10 liter bioreactor vessel. 316l stainless steel with electropolish finish 1/2-20 tpi threads For 10l bioreactor systems - English threads For 1/2" port on headplate Su
SDP
Supplier:  Bellco Glass, Inc. 196401012
Encompass_Preferred
Stainless steel port plug fits onto stainless steel head plate for 10 liter bioreactor vessel. 316l stainless steel with electropolish finish 1/2-20 tpi threads For 10l bioreactor systems - English threads For 1/2" port on headplate Su
Catalog No. 50-194-4715
Explore available promotions