Learn More
HOYA™ Super HiVision EX3 Coating
Catalog No. 18999585
Click to view available options
dimLensType: Anti-reflective
This item is not returnable. View return policy
This item is not returnable. View return policy
Provide twice the scratch resistance of any AR coating available while maximizing light to the eye to optomize sight. HOYA™ Super HiVision EX3 Coating creates a more durable lens with minimal glare.
  • Multi layer coating including AR scratch hydrophic properties
  • Offers the highest scratch resistance available
  • Ensures the highest level of light transmission
  • Provides more comfortable vision
  • Improved hydrophobic layer
  • Water and dirt repellent
  • Easy to clean, even after long term use and repeated wiping
Specifications
Lens Tint Clear
Product Type Add on - Lens Coating