Learn More
Supertek Scientific Pipet Pump
GREENER_CHOICESCI_ED
Fast release pipeting device for precise pipetting and zip quick emptying.
$11.50 - $13.50
Specifications
Type Pipette Pump
For Use With (Application) Precise pipetting and zip quick emptying
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
S09610B
GREENER_CHOICESCI_ED
View Documents
Supertek Scientific
A10120/3
10 mL
Each for $12.50
Add to Cart
S09610A
GREENER_CHOICESCI_ED
View Documents
Supertek Scientific
A10120/1
2 mL
Each for $11.50
Add to Cart
S09610C
GREENER_CHOICESCI_ED
View Documents
Supertek Scientific
A10120/5
25 mL
Each for $13.50
Add to Cart
Description
Description
  • Made from acrylonitrile butadiene styrene
  • With knurled thumbwheel that can be roated to draw up or dispense liquids
  • Push in releaser for zip-quick emptying
  • Silicon rubber collet inside moulded chuck holds pipets
  • Resistanct to acids and alkalies
  • Easily disassembled for cleaning purpose
Specifications
Specifications
Pipette Pump
Precise pipetting and zip quick emptying