Learn More
Tosoh Bioscience AIA-PACK™ Test Cups – CEA (Carcinoembryonic Antigen)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier:  Tosoh Bioscience 0025254
 View more versions of this product
Catalog No. 21-280-019
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions
Description
Description
  • These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
Specifications
Specifications
Carcinoembryonic Antigen
Serum
CEA Assay Test Cup
5 trays of 20 cups
100 μL
AIA Test Cup
0.5 ng/mL
Non-neoplastic and benign diseases such as Alcoholic cirrhosis, hepatitis, ulcerative colitis and Crohn's disease
Up to 100ng/mL