Learn More
Yamato Scientific America, Inc. Yamato 27L Vacuum Oven, Benchtop, 115V
Supplier:  Yamato Scientific America, Inc. ADP300C
Encompass_Preferred
Standard small size 27L benchtop vacuum drying oven with enhanced safety features. Easy input of parameters and settings. Digital PID controller supports fixed temperature, quick auto-stop, auto stop, auto start and program operations. Self-diagnostic and overheating prevention functions. Silicon rubber door seal prevents air from leaking. Independent over heating prevention device for each circuit. Customizable with N₂ gas inlet and communication ports. Calibration off-set function. Easy maintenance. Operating temp. range of 40~240°C *This unit requires a vacuum pump. If customer has one that meets the requirements of the oven they can use. If not, please search part number NC1949712*
Catalog No. 50-189-8031
Explore available promotions