Learn More
Saint Gobain Performance Plastics VAC TUBING 3/16 x 9/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACF00015
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Vacuum Sizes Diameter Inner 3/16 in. Diameter Outer 9/16 in. Thickness 3/16 in. Clear Maximum Pressure 69 psi at 73 deg.F
Catalog No. 50-206-8945
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Explore available promotions