Learn More
TWD Scientific LLC Vial, Cryovial, Twd Biox, 1.0Ml, 1D And 2D Barcoded, External Thread, White Cap, Sterile, Writing Patch, Marked Graduations, Polypropylene, 1000 Vials/Case
SDP
Supplier:  TWD Scientific LLC BX889100
Encompass_Preferred
1.0Ml Clear Polypropylene Sterile Pre-Set 1D & 2D Barcoded Cryovials With External Thread And White Caps Assembled. Cryovials Have Writing Patch And Marked Graduations. 25 Cryovials/Bag, 20 Bags/Pack, 2 Packs/Case, Price/Case
Catalog No. 50-228-5059
Explore available promotions