Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Wood Science Lab Table, Depth: 60 to 72 in.
SCI_ED

NPS Science Lab Tables have perfect chemistry with any science classroom.

Supplier:  National Public Seating™ SLT23072H

 View more versions of this product

Catalog No. S83652


Explore available promotions
Add to Cart

Description

Description

  • Solid ash legs and apron
  • Rubber liquid protecting boots
  • Legs attach with metal on metal double bolt Through assembly on 13-gauge steel corner braces
  • Adjustable glides for accurate leveling
  • 500 lb. weight capacity
  • 10 year warranty
Specifications

Specifications

Lab Table
Resists scratches, boiling water, high temperatures, and acid stains
Black
HPL
Science Labs
72 x 30 x 36 in.
MAS2, ANSI BIFMA
10 years
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.