Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Public Seating™ Wood Science Lab Table with Whiteboard Top
NPS Industrial, Fixed Height Whiteboard Top is an asset in any setting.
$700.00 - $1409.00
Specifications
Certifications/Compliance | MAS2, ANSI BIFMA |
---|---|
For Use With (Application) | Science Labs |
Top Style | Whiteboard |
Product Type | Lab Table |
Color | White |
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
S83626
|
National Public Seating™
SLT12448W |
48 x 24 x 30 in. |
Each for $700.00
|
|
|||||
S83654
|
National Public Seating™
SLT22448W |
48 x 24 x 36 in. |
Each for $866.00
|
|
|||||
S83627
|
National Public Seating™
SLT12454W |
54 x 24 x 30 in. |
Each for $762.00
|
|
|||||
S83655
|
National Public Seating™
SLT22454W |
54 x 24 x 36 in. |
Each for $911.00
|
|
|||||
S83628
|
National Public Seating™
SLT12460W |
60 x 24 x 30 in. |
Each for $795.00
|
|
|||||
S83656
|
National Public Seating™
SLT22460W |
60 x 14 x 36 in. |
Each for $962.00
|
|
|||||
S83629
|
National Public Seating™
SLT12472W |
72 x 24 x 30 in. |
Each for $880.00
|
|
|||||
S83657
|
National Public Seating™
SLT22472W |
72 x 24 x 36 in. |
Each for $1,034.00
|
|
|||||
S83630
|
National Public Seating™
SLT13060W |
60 x 30 x 30 in. |
Each for $893.00
|
|
|||||
S83658
|
National Public Seating™
SLT23060W |
60 x 30 x 36 in. |
Each for $1,071.00
|
|
|||||
S83631
|
National Public Seating™
SLT13072W |
72 x 30 x 30 in. |
Each for $987.00
|
|
|||||
S83659
|
National Public Seating™
SLT23072W |
72 x 30 x 36 in. |
Each for $1,136.00
|
|
|||||
S83632
|
National Public Seating™
SLT14260W |
60 x 42 x 30 in. |
Each for $1,288.00
|
|
|||||
S83660
|
National Public Seating™
SLT24260W |
60 x 42 x 36 in. |
Each for $1,409.00
|
|
|||||
Description
- Whiteboard is made from 1.25 in. industrial grade particleboard with a high-pressure laminate top and 3 mm edge banding
- Solid ash legs and apron
- Rubber liquid protecting boots
- Legs attach with metal on metal double bolt through assembly on 13-gauge steel corner braces
- Adjustable glides for accurate leveling
- 10 year warranty
Specifications
MAS2, ANSI BIFMA | |
Whiteboard | |
White |
Science Labs | |
Lab Table |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title