Learn More
Electron Microscopy Sciences Write on Marker Pens, 10/PK
Supplier: Electron Microscopy Sciences 7210610A
These pens are made from specially formulated ink and are to be used on anterior surfaces as well as sides of histology cassettes. The ink will dry almost instantly and withstands temperatures up to 195 degree C and is resistant to solvents.