Learn More
StatLab ZAMBONI`S FIXATIVE /PINT
SDP
Supplier:  StatLab FXZAMPT
Encompass_Preferred
Excellent for ER or PR immunostains and Electron Microscopy. Zamboni’s stabilizes cellular proteins, is not easily destroyed by tissue fluids, and will not cause tissue deterioration. This allows specimens to be kept at room temperature indefinitely and be sent through the mail without the surgical samples being compromised. Prefilled Specimen Containers are pre-labeled, leak-tight, and filled to 50% of the container's capacity.
Catalog No. NC0200279
Explore available promotions