Learn More
TARGETMOL CHEMICALS INC ZD 7155 HYDROCHLORIDE 5MG
Supplier: TARGETMOL CHEMICALS INC T13390
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZD 7155 hydrochloride is an angiotensin II receptor type 1 (AT1 receptor) antagonist. purity: 99%