Learn More
Zymo Research Corporation ZR-96 Zymoclean™ Gel DNA Recovery Kit (2 x 96 Preps)
Supplier: Zymo Research Corporation D4021
Simply add the specially formulated Agarose Dissolving Buffer (ADB) to the gel slices containing DNA, let dissolve, and then transfer to the wells of the supplied ZymoSpin™ I-96 Plate. There is no need for organic denaturants or chloroform.Instead, the product utilizes Fast-Spin technology to yield high-quality DNA in just minutes. DNA purified using the ZR-96 Zymoclean™ Gel DNA Recovery Kit is well suited for DNA ligation, sequencing, DNA labeling, PCR, etc.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.