Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Oxa-6-azaspiro[3.3]heptane hemioxalate, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
500 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 500 mg |
IUPAC Name | bis(2-oxa-6-azaspiro[3.3]heptane); oxalic acid |
Formula Weight | 144.15 |
Percent Purity | 96% |
Chemical Name or Material | 2-Oxa-6-azaspiro[3.3]heptane hemioxalate |
Safety and Handling
Hazard Category | H302+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H312 Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Keep cold |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction