Learn More
Assay Technology 566 ORGANIC VAPOR MONITORS 5PK
Supplier: Assay Technology X5664AT
The 566 Organic Vapor Monitor is our most versatile and popular organic vapor badge. The 566 utilizes our ChemDisk II configuration with a flexible inert charcoal wafer making it smaller and much easier to use than other badges on the market. The 76-hole sampling grid allows for sampling rates around 8 mL/min.