Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Braze Composite Safety Toe
A classic broad toe oxford that provides plenty of toe room and a composite safety toe cap that protects your feet. Its sharp, rugged looks make it a great option for work, and for everyday wear.
Specifications
Color | Brown |
---|---|
Material | Leather |
Sole Material | Rubber |
Toe Material | Composite |
Catalog Number | Mfr. No. | Size | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Width (English) | Price | Quantity & Availability | ||||
19-154-262
|
Carolina™
CA15200802E |
8 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-273
|
Carolina™
CA1520080D |
8 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-263
|
Carolina™
CA15200852E |
8.5 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-274
|
Carolina™
CA1520085D |
8.5 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-264
|
Carolina™
CA15200902E |
9 | 2E | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-154-275
|
Carolina™
CA1520090D |
9 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-265
|
Carolina™
CA15200952E |
9.5 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-276
|
Carolina™
CA1520095D |
9.5 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-266
|
Carolina™
CA15201002E |
10 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-277
|
Carolina™
CA1520100D |
10 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-267
|
Carolina™
CA15201052E |
10.5 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-278
|
Carolina™
CA1520105D |
10.5 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-268
|
Carolina™
CA15201102E |
11 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-279
|
Carolina™
CA1520110D |
11 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-269
|
Carolina™
CA15201152E |
11.5 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-280
|
Carolina™
CA1520115D |
11.5 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-270
|
Carolina™
CA15201202E |
12 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-281
|
Carolina™
CA1520120D |
12 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-271
|
Carolina™
CA15201302E |
13 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-282
|
Carolina™
CA1520130D |
13 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-272
|
Carolina™
CA15201402E |
14 | 2E | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-154-283
|
Carolina™
CA1520140D |
14 | D | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Brown
- Full Grain Leather
- Composite Safety Toe
- ESD
- Non-Metallice
- Oxford
Specifications
Brown | |
Leather | |
Rubber | |
Composite |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title