Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Lightweight Nomex™ Flame Resistant Uniform Shirt
Click to view available options
Color:
Navy
Royal Blue
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
Description
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 1 Protection
- Protection: Arc Rating ATPV 4.8 calories/cm2
- Hazard: 1
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 4.5 oz. (150 g/m2) Nomex™ IIIA
- Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
- Closure: Button-down front and topstitched cuff with button closure
- Collar: Banded topstitched collar
- Pocket: Two chest pockets with button flap closures and sewn in pocket stall
- Placket front with button closure
- Tailored sleeve placket
- Product Collection: Nomex™
- Protection: ATPV Arc Rating: 4.4
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Nomex™
- Brand: Bulwark™
Specifications
Specifications
Size | Small (Regular) |
Gender | Men's |
Material | Nomex IIIA |
Color | Navy |
Closure Type | Button |
Hazard Risk Category | 1 |
Arc Flash Rating | ATPV 4.8 Calories/cm2 |
Certifications/Compliance | NFPA 2112, NFPA 70E, Category 1 Protection |
Weight (English) | 4.5 oz. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction